missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Tryptic Soy Agar

Catalog No. ICN1010617
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1010617 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1010617 Supplier MP Biomedicals™ Supplier No. 091010617

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for the cultivation of fastidious and non-fastidious microorganisms.

Tryptic Soy Agar is a medium very rich in nutrients for 'general use' in microbiological laboratories.

Containing two peptones obtained by enzymatic hydrolysis of casein and soy protein, this medium supports the growth of a great variety of microorganisms, including fastidious aerobes and anaerobes.

If desired, antibiotics can easily be incorporated as well as other supplements or inhibitory agents.

A short list of microorganisms that grow on this medium are the following: Streptococcus, Neisseria, Brucella, Corynebacterium, Listeria, Pasteurella, Vibrio, Haemophilus vaginals, Candida, etc.

Specifications

Content And Storage 15 to 30°C
Form Powder
Media Type Tryptic Soy Agar
Product Type Tryptic Soy Agar
Formulation Casein Peptone 15 g/L, Sodium Chloride 5 g/L, Soy Peptone 5 g/L, Agar 15 g/L
For Use With (Application) Isolation and cultivation of fastidious organisms
Quantity 500 g

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.