missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ TeenFlex™ Adapter for 20 x 15 mL Tube Holder on FastPrep 96
Description
- Applications: TeenFlex™ 20 x 15mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing and forensic applications.
- Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Product Overview: With the FastPrep-96 Instrument, TeenFlex adapter and optimized Lysing Matrices, consistent reproducible results are obtained.
- Product Description: Medium-scale adapter for the FastPrep-96™ that holds 20 x 15 mL Lysing Matrix Tubes (compatible with SafTest™ Food Inspection System.) It allows frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes. A wide range of disposable 15 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.
- Recommended Use: Use with FastPrep-96, MP Bio Lysing Matrix Tubes and/or FastPrep purification kits to lyse, grind or homogenize samples at ambient temperatures, for the extraction and purification of nucleic acids, proteins, metabolites and other small molecules and more.
- Capacity: 20 tubes
- Sample Volume: Up to 40 g
Specifications
Specifications
| Product Type | FastPrep-96 Adapters |
| For Use With (Application) | Extractions of nucleic acids, proteins, metabolites and other small molecule |
| For Use With (Equipment) | FastPrep-96 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.