missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TallPrep™ Adapter for 24 x 4.5 mL Tube Holder on FastPrep-24

TallPrep™ Adapter for 24 x 4.5 mL Tube Holder on FastPrep-24 is used for room temperature lysis and grinding of samples.

Supplier:  MP Biomedicals™ 116002540

Catalog No. MP116002540

Only null left

Description

Description

TallPrep™ adapter holds 24 x 4.5 mL Lysing Matrix tubes. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix tubes that remain securely sealed during the homogenization. It stands stable on the bench top and is commonly used as a tube rack. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.

  • Application: TallPrep™ 24 X 4.5 mL Adapter is ideally suited for high-throughput applications, in which up to 24 samples can be homogenized simultaneously.
  • Capacity: 20 tubes
  • Sample Volume: Up to 10 g
Specifications

Specifications

Adapter
FastPrep-24 and FastPrep-96
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.