missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TallFlex™ Adapter for 48 x 4.5 mL Tube Holder on FastPrep 96

TallFlex™ Adapter for 48 x 4.5 mL Tube Holder on FastPrep 96 is used for room temperature grinding.

Supplier:  MP Biomedicals™ 116010580

Catalog No. MP116010580


Only null left

Description

Description

  • Product Overview: Medium-scale adapter for the FastPrep-96™ that holds 48 x 4.5 mL Lysing Matrix Tubes. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix tubes that remain securely sealed during the homogenization. It stands stable on the bench top and is commonly used as a tube rack. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Application: TallFlex™ 48 x 4.5 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Capacity: 48 tubes
  • Sample Volume: Up to 20 g
Specifications

Specifications

FastPrep-96 Adapters
Room Temperature
Extractions of nucleic acids, proteins, metabolites and other small molecules from samples
48 x 4.5 mL Tube Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.