missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SD MEDIUM-URA, Powder
Complete synthetic defined single dropout medium for the growth of S. cereviseae
Supplier: MP Biomedicals™ 114813065
Description
Yeast cell culture, Gene expression, Transcription regulation
Specifications
| Molecular biology Certified™ | |
| Powder | |
| YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L, CSM-Ura 0.69g/L | |
| Pouch |
| 15°C to 30°C | |
| Yeast Media | |
| For growth of Saccharomyces cerevisiae | |
| 10 x 500 mL |