missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RPMI 1640 Medium, 1X Liq., w/o L-Glutamine and Glucose

Catalog No. ICN1646854
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1646854 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1646854 Supplier MP Biomedicals™ Supplier No. 091646854

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas.

Specifications

Product Type RPMI 1640
Sterility Sterile
Without Additives No Glutamine, No Glucose
Quantity 500 mL
pH 6.8 to 7.7
Content And Storage 4°C