missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ready to Use 5% 7X™ Solutions
GREENER_CHOICE

Catalog No. ICN7668193
Change view
Click to view available options
Description:
7X-O-Matic; 1 gal.
7X; 1 gal.
ES 7X; 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
ICN7668193 ES 7X; 1 gal.
ICN7668093 7X; 1 gal.
ICN7668493 7X-O-Matic; 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN7668193 Supplier MP Biomedicals™ Supplier No. 097668193

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing

  • Ideal for glass and plastic labware
  • Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
  • Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds

Specifications

Form Liquid
Concentration 7X
Content And Storage 15°C to 30°C
Description ES 7X; 1 gal.
For Use With (Application) For Glass, Plastic, benchtop equipment and tubing
Volume (English) 1 gal.