missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Protamine Sulfate
Description
Cell Culture Reagent. Highly purified. Nitrogen: Approx. 22.5%. Hygroscopic white powder.
Specifications
Specifications
| Content And Storage | Room Temperature |
| Format | Powder |
| For Use With (Application) | Antiheparin activity, refining of vaccines, removal of DNA from Protein samples |
| Product Type | Protamine Sulfate from Salmon Sperm |
| Quantity | 5000 mg |