missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Plant Culture Salt Mixtures

Catalog No. ICN2633024
Change view
Click to view available options
Quantity:
10 x 1 L
1 x 10 L
1 x 50 L
Product Type:
Hoagland's Modified Basal Salt Mixture
Murashige and Skoog Basal Salt Mixture
Murashige and Skoog Plant Salt Mixture
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ICN2633024 1 x 50 L Murashige and Skoog Plant Salt Mixture
ICN2621820 10 x 1 L Hoagland's Modified Basal Salt Mixture
ICN2621822 1 x 10 L Hoagland's Modified Basal Salt Mixture
ICN2623020 10 x 1 L Murashige and Skoog Basal Salt Mixture
ICN2623022 1 x 10 L Murashige and Skoog Basal Salt Mixture
ICN2633022 1 x 10 L Murashige and Skoog Plant Salt Mixture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN2633024 Supplier MP Biomedicals™ Supplier No. 092633024
Only null left

Plant Culture Salt Mixtures are for the molecular testing of plants and plant-derived samples for improving the productivity, quality, and disease resistance of plants.

  • Ready to use
  • Liquid formulation, prepared according to the original specification, except where noted
  • Available in different pack sizes, ready to use and liquid formulations

Specifications

Form Powder
Product Type Murashige and Skoog Plant Salt Mixture
Without Additives No Agar
Quantity 1 x 50 L
Format Solution
pH 4.8 to 6.2
Content And Storage 4°C