missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Multitest Slides

Catalog No. ICN6041805
Change view
Click to view available options
Wells:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
ICN6041805 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN6041805 Supplier MP Biomedicals™ Supplier No. 096041805

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

10-well slides with many applications including immunofluorescense, cell culture, slide agglutination tests and histology

The uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.

  • Material: Fragile glass with blue PTFE coating
  • Dimensions: 8mm (0.31 in.) well diameter arranged 2 x 5
  • Average size: 2.5cm (0.98 in.) × 7.5cm (2.95 in.) × 1mm (0.04 in.)

  • A 4-channel pipette can be used to transfer samples directly from the microplate to the slide
  • Not a reusable slide - soaking and washing after use can cause the PTFE coating to lift off due to previous reactions of organic solvents and fixatives

Specifications

Type Multitest Slide
Wells 10
Material Glass
For Use With (Application) Immunofluorescense, cell culture, slide agglutination tests and histology
Dimensions (LxW) 25 x 75 mm