missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Linbro™ Microplate Sealer
Specifications
Specifications
| Color | Clear |
| Material | Mylar |
| No. per Pack | 100 |
| Product Type | Microplate Sealer |
| Color | Clear |
| Material | Mylar |
| No. per Pack | 100 |
| Product Type | Microplate Sealer |