missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Linbro™ Microplate Sealer

Catalog No. MP0976401C2
Change view
Click to view available options
No. per Pack:
100
1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
MP0976401C2 1000
ICN7640105 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP0976401C2 Supplier MP Biomedicals™ Supplier No. 0976401C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with multi-well plates and tissue culture plates

  • Easy application and removal for all microplates for both short- and long-term storage Provide tight seal to minimize evaporation and condensation
  • Suitable for use between -16 °C and 38 °C
  • Transparent tape and is not pierceable
  • Nonsterile

Specifications

Color Clear
Material Acetate
Sterility Non-sterile
No. per Pack 1000
For Use With (Application) Used for multi-well plates and tissue culture plates
Product Type Microplate Sealer
Temperature Range (Metric) -16°C to +38°C