missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LB-Agar, Powdered

Catalog No. MP113002232
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
Format:
Capsule
Powder
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
MP113002232 1000 g Powder
MP113002222 454 g Powder
MP113002231 1000 g Capsule
MP113002241 5000 g Capsule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP113002232 Supplier MP Biomedicals™ Supplier No. 113002232
Only null left

Offered in a variety of formats for convenience and ease of use

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Format Powder
Grade Molecular biology
Product Type Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application) For growth of bacteria
Quantity 1000 g