missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ham's F-10 Medium

Catalog No. MP091440049
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091440049 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091440049 Supplier MP Biomedicals™ Supplier No. 091440049

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized for clonal isolation and growth of CHO cells

Specifications

Product Type Ham's F-10 Medium
Sterility Sterile
Without Additives No Glutamine, No Sodium Bicarbonate
Quantity 100 mL
pH 5.5 to 6.1
Content And Storage 4°C