missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ginsenoside RG1

Catalog No. ICN19889105
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19889105 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19889105 Supplier MP Biomedicals™ Supplier No. 0219889105
Only null left

From Ginseng root

Reference grade. Purity: ∽95%.

Ref.: Shibata, S., et al., Pharmacol. Bull, 14: 595, (1966).

Specifications

Purity 98%
Content And Storage 15°C to 30°C
Format Powder
Molecular Weight (g/mol) 801.04
Product Type Ginsenoside RG1 from Ginseng Root
Quantity 5 mg