missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fibronectin, 1mg
Specifications
Specifications
| Concentration | 5.3mg/mL |
| For Use With (Application) | Cell Culture, Immunoassay |
| Name | Fibronectin from Human Donor Plasma |
| pH Range | 7.3 |
| Preparation Method | Human Fibronectin is purified from Human Plasma using multi-step procedures |
| Purification Method | Gel Filtration/Ion Exchange Chromatography, Immunoabsorption |
| Quantity | 1 mg |
| Source | Human Donor Plasma |
| Storage Requirements | 2° to 8°C |