missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ham's F-12 (Modified) with L-glutamine, without Sodium Bicarbonate

Catalog No. MP091042122
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091042122 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091042122 Supplier MP Biomedicals™ Supplier No. 091042122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Well suited for the growth of primary rat hepatocytes and prostate epithelial cells.

Ham's F-12 medium was designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells. Ham's F-12 is well suited for the growth of primary rat hepatocytes and prostate epithelial cells. It is also the medium of choice for toxicity assays using CHO cells.

Specifications

Form Powder
Product Type Nutrient Medium
With Additives Glutamine
Without Additives No Sodium Bicarbonate
Quantity 1 x 10 L
pH 6.3 to 7.2
Content And Storage 0 to 5°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.