missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ DMEM/Ham's F-12 Medium without L-glutamine, sodium bicarbonate

Catalog No. MP091046822
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091046822 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091046822 Supplier MP Biomedicals™ Supplier No. 091046822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powder; With 3151 mg/L dextrose

  • Acts as mammalian culture media
  • Used as basal media for serum-free cell culture

Specifications

Form Powder
Product Type Dulbecco's Modified Eagle Medium
With Additives Dextrose
Without Additives No Glutamine, No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture applications
Quantity 1 x 10 L
pH 7.6
Content And Storage 0 to 5°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.