missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Corn Syrup

Catalog No. ICN10141301
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10141301 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10141301 Supplier MP Biomedicals™ Supplier No. 0210141301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dual acid-enzyme conversion syrup

Contains three times the maltose of regular corn syrup.

  • Liquid
  • Maltose content approximately 30%
  • Processed using acid only

Specifications

Product Type Animal Diet
Format Liquid
For Use With (Application) Corn syrup are used in commercially prepared foods as a thickener, a sweetener and as a humectant - an ingredient that retains moisture and thus maintains a food's freshness
Quantity 1 gal.
Storage Requirements 15°C to 30°C