missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CoolBigPrep™ Adapter for 2 x 50 mL Tube Holder on FastPrep-24

CoolBigPrep™ Adapter on FastPrep-24 holds 2 x 50 mL tubes for cryogenic cell grinding and lysis. It is used for extraction of sensitive biological compounds.

Supplier:  MP Biomedicals™ 116002531

Catalog No. MP116002531


Only null left

Description

Description

  • Product Overview: CoolBigPrep™ 2 x 50 mL Adapter is a cryogenic adapter which allows simultaneous lysis of 2 x 50 mL samples. Based on passive temperature control technology, the CoolBigPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep™ precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. A wide range of disposable 50 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in 40 seconds or less.
  • Applications: CoolBigPrep™ 2 x 50 mL Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
  • Capacity: 2 tubes
  • Sample Volume: Up to 40 g
Specifications

Specifications

Up to 40 g
Ideal for extractions of temperature unstable biological compounds such as RNA, siRNA, metabolites, intermediates and enzymes
FastPrep™- 24, FastPrep™-96
2 x 50 mL Samples
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.