missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Brewers Yeast

Used as an insect diet.

Supplier:  MP Biomedicals™ 0290331280

 View more versions of this product

Catalog No. ICN90331280

Only null left
Explore available promotions

Description

Description

  • Source of B-complex vitamins, chromium and selenium
  • Brewers Yeast is commonly used for baking break, fermenting wine and for beer and ethanol production
Specifications

Specifications

Animal Diet
Source of B-complex Vitamins, Chromium and Selenium, also used to bake bread, ferment wine, beer and Ethanol production
S. cerevisiae
Powder
45.35 kg
15°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions