missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ BioRich 1
Description
- DMEM/F-12 powdered media blend enriched with trace elements
- Permits the transition to serum-free conditions, reducing or eliminating the necessity for serum supplementation
- Use with ITS™ Premix, a completely serum-independent universal supplement
Specifications
Specifications
| Product Type | DMEM/F-12 Medium |
| Without Additives | No Sodium Bicarbonate |
| Quantity | 1 x 10 L |
| pH | 7.1 to 7.7 |
| Content And Storage | 4°C |