missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ BigFlex™ Adapter for 8 x 50 mL Tube Holder for FastPrep 96

Catalog No. MP116010550
Change view
Click to view available options
For Use With (Equipment):
FastPrep-96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
MP116010550 FastPrep-96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116010550 Supplier MP Biomedicals™ Supplier No. 116010550

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BigFlex™ Adapter for 8 x 50 mL Tube Holder for FastPrep is a large sample volume adapter suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications.

  • Product Overview: BigFlex™ 8 x 50 mL Adapter for FastPrep-96™ is a large-scale adapter for the FastPrep-96™ that holds 8 x 50 mL Lysing Matrix tubes (compatible with SafTest™ Food Inspection System). A wide range of disposable 50 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.
  • Application: BigFlex™ 8 x 50 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Capacity: 8 tubes
  • Sample Volume: Up to 160 g

Specifications

Sample Volume (Metric) Up to 160 g
For Use With (Application) To grind or homogenize samples at ambient temperature
For Use With (Equipment) FastPrep-96
Holds 8 x 50 mL Tubes

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.