missing translation for 'onlineSavingsMsg'
Learn More

Moxonidine 98.0+%, TCI America™

Catalog No. p-7133355
Change view
Click to view available options
Quantity:
1 g
200 mg
Catalog No. Quantity
M2660200MG 200 mg
M26601G 1 g
2 options

Catalog No. M2660200MG

Supplier: TCI America M2660200MG

Only null left

Chemical Identifiers

CAS 75438-57-2
Molecular Formula C9H12ClN5O
Molecular Weight (g/mol) 241.68
MDL Number MFCD22689455
InChI Key WPNJAUFVNXKLIM-UHFFFAOYSA-N
Synonym 4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine
PubChem CID 4810
ChEBI CHEBI:7009
IUPAC Name 4-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methylpyrimidin-5-amine
SMILES COC1=NC(C)=NC(Cl)=C1NC1=NCCN1

Specifications

Melting Point 206°C
Color White
Quantity 200 mg
UN Number 2811
Formula Weight 241.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Moxonidine
RTECSNumber UV6260290
TSCA No