missing translation for 'onlineSavingsMsg'
Learn More

Moxifloxacin hydrochloride, 97%

Catalog No. p-7057521
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66626MC 100 mg
AAJ66626ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66626MC Supplier Thermo Scientific Chemicals Supplier No. J66626MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 186826-86-8 | C21H25ClFN3O4 | 437.896 g/mol

Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

Solubility
Sparingly soluble in water. Soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 186826-86-8
Molecular Formula C21H25ClFN3O4
Molecular Weight (g/mol) 437.896
MDL Number MFCD00949117
InChI Key IDIIJJHBXUESQI-DFIJPDEKSA-N
PubChem CID 101526
ChEBI CHEBI:7008
IUPAC Name 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl

Specifications

Type Moxifloxacin Hydrochloride
Physical Form Powder
Odor Odorless
Color Yellow
Quantity 100 mg
Flash Point >110°C (230°F)
Boiling Point 636°C
Beilstein 8377447
Solubility Information Sparingly soluble in water; Soluble in DMSO
Formula Weight 437.89
Percent Purity 97%
Chemical Name or Material Moxifloxacin hydrochloride
Show More Show Less
Hazard Category H302-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
RTECSNumber VB1983750
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only