missing translation for 'onlineSavingsMsg'
Learn More

Moxalactam sodium salt

Catalog No. AC460410010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460410010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460410010 Supplier Thermo Scientific Chemicals Supplier No. 460410010
Only null left

CAS: 64953-12-4 | C20H18N6Na2O9S | 564.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64953-12-4
Molecular Formula C20H18N6Na2O9S
Molecular Weight (g/mol) 564.44
InChI Key GRIXGZQULWMCLU-UHFFFAOYNA-L
IUPAC Name disodium 7-[2-carboxylato-2-(4-hydroxyphenyl)acetamido]-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].[Na+].COC1(NC(=O)C(C([O-])=O)C2=CC=C(O)C=C2)C2OCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 564.44
Chemical Name or Material Moxalactam sodium salt
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction.

May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

In case of inadequate ventilation wear respiratory protection.

IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

EINECSNumber 265-288-4

RUO – Research Use Only