Learn More
Kent Scientific MouseSTAT™ Pulse Oximeter Whole Body Sensor for Mouse and Pup
For use with PhysioSuite and PhysioSuite Modules.
$1,049.05 - $1,772.63
Specifications
Animal Mouse and pups that are less than 3 days old
For Use With (Application) Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring
For Use With (Equipment) PhysioSuite and PhysioSuite Modules
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
13005135
Encompass
View Documents Promotion Details
Kent Scientific
SPO29MM
Pulse Oximeter Whole Body
Each for $1,049.05
Only null left
13005136
Encompass
View Documents Promotion Details
Kent Scientific
SPO29MMMRI
Pulse Oximeter Whole Body/MRI Comp.
Each for $1,772.63
Only null left
Description
Description
  • Recommended for use with mouse and pups that are less than 3 days old
  • MRI compatible sensor includes 3m cable
Applications
Vital sign monitoring, pulse oximetry, heart rate, physiological monitoring
Specifications
Specifications
Mouse and pups that are less than 3 days old
Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring
PhysioSuite and PhysioSuite Modules