Learn More
Kent Scientific MouseSTAT™ Pulse Oximeter Whole Body Sensor
Catalog No. 13005137
Encompass
Change view
Click to view available options
Type:
Pulse Oximeter Whole Body
Pulse Oximeter Whole Body/MRI Comp.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13005137 Pulse Oximeter Whole Body
13005138 Pulse Oximeter Whole Body/MRI Comp.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13005137 Supplier Kent Scientific Supplier No. SPO213MM
For use with PhysioSuite and PhysioSuite Modules.
  • Recommended for use with mouse and pups that are greater than 3 days old
  • MRI compatible sensor includes 3m cable
Applications
Vital sign monitoring, pulse oximetry, heart rate, physiological monitoring
Specifications
Animal Mouse and pups that are greater than 3 days old
For Use With (Application) Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring
Type Pulse Oximeter Whole Body
For Use With (Equipment) PhysioSuite and PhysioSuite Modules