Learn More
Acrobiosystems ACROBiosystems MouseAnti-SARS-CoV-2AntibodyIgGTiterSerologicAssaykit
Supplier: ACROBiosystems Inc RAST01896TESTS
Kits;96TESTS;This assay kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in Mouse serum by SARS-CoV-2 Spike RBD. The Kit consists of Pre-coated with SARS-CoV-2 Spike RBD Microplate, an Anti-SARS-CoV-2 Antibody (Control, Mouse IgG1), an HRP-Anti-Goat anti mouse IgG secondary antibody.;Detection of Mouse Anti-SARS-CoV-2 Antibody IgG titer by Indirect-ELISA Assay.
Immobilized SARS-CoV-2 Spike RBD at 2 g/mL (100 L/well) can bind Mouse Anti-SARS-CoV-2 Antibody IgG in mouse serum. Detection was performed using HRP-Conjugated Anti-mouse IgG secondary antibody with sensitivity of 20 ng/mL (QC tested).