missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse VCP (aa 792-806) Synthetic Peptide

Catalog No. PEP239
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP239 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP239 Supplier Invitrogen™ Supplier No. PEP239

Synthetic Peptide

Mouse VCP (residues 792-806) Synthetic Peptide, PEP-239, from Invitrogen.

The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.

Specifications

Gene ID (Entrez) 269523
Species Mouse
Purification Method purified
Accession Number Q01853
Common Name Mouse VCP (aa 792-806)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Vcp
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C G(792) G S V Y T E D N D D D L Y G(806)
Show More Show Less