missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Mouse STEMCCA Cre-Excisable Constitutive Polycistronic (OKS) Lentivirus Reprogramming Kit

Catalog No. SCR518
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR518 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR518 Supplier MilliporeSigma™ Supplier No. SCR518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains high titer Cre-excisable polycistronic (OKS) lentivirus that expresses modified stem cell cassette. MilliporeSigma™; Mouse STEMCCA Cre-Excisable Constitutive Polycistronic (OKS) Lentivirus Reprogramming Kit comprises of the three transcription factors Oct4, Klf4, and Sox2 (OKS) separated by the self-cleaving 2A peptide and IRES sequences.

  • Components:
  • Mouse STEMCCA Constitutive LoxP (OKS) Lentivirus: Two (2) vials of EF1α-STEMCCA-LoxP (OKS) Lentivirus (Part No. CS204448). Each vial contains 15μL of high titer lentivirus
  • TGF-ß RI Kinase Inhibitor II Supplement (1000X): (Part No. CS204458) One (1) vial containing 300μL of the inhibitor in high quality DMSO
  • GSK3ß Inhibitor Supplement (1000X): (Part No. CS204418) One (1) vial containing 300μL of the inhibitor in high quality DMSO
  • Methylation Modulator-A Supplement (1000X): (Part No. CS204419) One (1) vial containing 300μL of the inhibitor in water
  • Polybrene 10 mg/mL: (Part No. TR-1003-50UL) One (1) vial containing 50μL of 10mg/mL stock of Polybrene transfection reagent

Specifications

Product Type Lentivirus Reprogramming Kit
For Use With (Application) Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage Store at -20°C.