missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse S1P1 (aa 359-372) Synthetic Peptide

Catalog No. PEP220
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP220 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP220 Supplier Invitrogen™ Supplier No. PEP220

Synthetic Peptide

Hyuman S1P1 (residues 359-372) Synthetic Peptide, PEP-220, from Invitrogen.

S1P1 is a G-protein coupled receptor for the bioactive lysosphingolipid sphingosine 1-phosphate (S1P) that is coupled to the G(i) subclass of heteromeric G proteins. S1P1 signaling leads to the activation of RAC1, SRC, PTK2/FAK1 and MAP kinases. It plays an important role in cell migration via its role in the reorganization of the actin cytoskeleton and formation of lamellipodia in response to stimuli that increase the activity of the sphingosine kinase SPHK1. S1P1 is required for normal chemotaxis toward sphingosine 1-phosphate, normal embryonic heart development, and normal cardiac morphogenesis.

Specifications

Gene ID (Entrez) 1901
Species Mouse
Purification Method purified
Accession Number P21453
Common Name Human S1P1 (aa 359-372)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol S1PR1
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence SHPQKDDGDN PETI
Show More Show Less