missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Regulatory T Cell Staining Kit #2

Catalog No. 501129225
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129225 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide and formaldehyde
Catalog No. 501129225 Supplier Invitrogen™ Supplier No. 88811840

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide and formaldehyde

Includes

  • Anti-Mouse CD4 FITC (RM4-5): 50 μg. Use at 0.125 μg/test
  • Anti-Mouse CD25 PE (PC61.5): 50 μg. Use at 0.06 μg/test
  • Anti-Mouse/Rat Foxp3 APC (FJK-16s): 25 μg. Use at 0.5 μg/test
  • Rat IgG2a K Isotype Control APC: 50 μg. Use at 0.5 μg/test
  • Anti-Mouse CD16/32 Purified: 50 μg Use at 2-5 μg/test.
  • Flow Cytometry Staining Buffer
  • Fixation/Permeabilization Concentrate: 30mL

Formulated & optimized for the staining of mouse splenocytes with the FJK-16s monoclonal antibody. eBioscience™ Mouse Regulatory T Cell Staining Kit #2 includes: Anti-Mouse CD4 FITC, CD25 PE and CD16/32, Anti-Mouse/Rat Foxp3 APC, Rat IgG2a Isotype Control APC, staining buffer, fixation concentrate.

Kit Contents:

  • Anti-Mouse CD4 FITC (RM4-5): 50μg—Use at 0.125μg/test
  • Anti-Mouse CD25 PE (PC61.5): 50μg—Use at 0.06μg/test
  • Anti-Mouse/Rat Foxp3 APC (FJK-16s): 25μg—Use at 0.5μg/test
  • Rat IgG2a K Isotype Control APC: 50μg—Use at 0.5μg/test
  • Anti-Mouse CD16/32 Purified: 50μg—Use at 2-5μg/test
  • Flow Cytometry Staining Buffer
  • Fixation/Permeabilization Concentrate: 30mL
  • Not included: Isotype controls for anti-CD4 (rat IgG2a, Cat. No. 11-4321) and anti-CD25 (rat IgG1, Cat. No. 12-4301)

The FJK-16s antibody reacts with mouse/rat Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3 a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human.

Intracellular staining of mouse splenocytes with FJK-16s using the mouse Foxp3 staining sets and protocol reveals approximately 2% of total splenocytes in the C57Bl/6 strain and approximately 3-5% in the BALB/c mouse strain. Multicolor flow cytometric analysis demonstrates approximately 90% of the CD4+CD25+ cells and 4% of the CD4+CD25- cells staining with FJK-16s. B220+, CD11b+, CD11c+, and Ly6G/Gr-1+ cells do not show significant co-staining with FJK-16s. These data are consistent with a recent report which follows expression of Foxp3 using a GFP knock-in (Fontenot et al, 2005).

FJK-16s cross-reacts with rat Foxp3 This has been demonstrated by intracellular staining of Foxp3 and flow cytometry of rat splenocytes using the same method and reagents as used for mouse tissue. Please note that the CD4 and CD25 antibodies included in this kit only recognize the mouse antigens. For staining rat tissue, please use (CD4 FITC, Cat. No. 11-0040 and CD25 PE, Cat. No. 12-0390).

Specifications

Content And Storage 2° to 8°C. Light sensitive material.
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 1 kit
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eBioscience
Product Type Cell Staining Kit

For Research Use Only. Not for use in diagnostic procedures.