missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse RARB (aa 429-448) Synthetic Peptide

Catalog No. PEP005
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP005 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP005 Supplier Invitrogen™ Supplier No. PEP005
Only null left

Synthetic Peptide

Mouse Retinoic Acid (residues 429-448) Synthetic Peptide, PEP-005, from Invitrogen.

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.

Specifications

Gene ID (Entrez) 218772
Species Mouse
Purification Method purified
Accession Number P22605
Common Name Mouse RARB (aa 429-448)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Rarb
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence P S V S P S S V E N S G V S Q S P L L Q
Show More Show Less