missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse PDE6B (aa 20-36) Synthetic Peptide

Catalog No. PEP183
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP183 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP183 Supplier Invitrogen™ Supplier No. PEP183

Synthetic Peptide

Mouse PDE6 beta (residues 20-36) Synthetic Peptide, PEP-183, from Invitrogen.

The second messengers cAMP and cGMP are key regulatory molecules that are involved in a wide variety of signal transduction pathways. Levels of cAMP and cGMP are regulated by their rate of synthesis by nucleotide cyclases and by their rate of hydrolysis by cyclic nucleotide phosphodiesterases (PDEs). PDEs form a superfamily of enzymes that catalyze the conversion of 3-prime, 5-prime-cyclic nucleotides to the corresponding nucleoside 5-prime-monophosphates. PDE6 is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha' (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). The catalytic core of the PDE6 system is comprised of alpha'/alpha' homodimers in the cone and alpha/beta heterodimers in the rod. The C-terminus of both the catalytic and inhibitory subunits is modified by methylation, myristyolation and prenylation which have been shown to be critical for proper complex assembly and membrane association.

Specifications

Gene ID (Entrez) 18587
Species Mouse
Purification Method purified
Accession Number P23440
Common Name Mouse PDE6B (aa 20-36)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Pde6b
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence H(20)-Q-Y-F-G-(K/R)-K-L-S-P-E-N-V-A-G-A-C(36)
Show More Show Less