missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse PCSK2 (aa 622-638) Synthetic Peptide

Catalog No. PEP138
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP138 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP138 Supplier Invitrogen™ Supplier No. PEP138
Only null left

Synthetic Peptide

Mouse PC2 (residues 622-638) Synthetic Peptide, PEP-138, from Invitrogen.

The subtilisin-like prohormone convertase (PC) family is a group of cellular enzymes that cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. PC family members share structural similarities, which include a heterogeneous approximately 10 kDa amino-terminal proregion, a highly conserved approximately 55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. The subcellular localization of PC family members varies. Immunolocalization studies show that PC1 is found in the perinuclear region as well as the trans-Golgi network, whereas PC2 can be found in the trans-Golgi network as well as diffusely distributed in the peripheral cytoplasm.

Specifications

Gene ID (Entrez) 18549
Species Mouse
Purification Method purified
Accession Number P21661
Common Name Mouse PCSK2 (aa 622-638)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Pcsk2
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence E(622)-L-D-E-A-V-E-R-S-L-Q-S-I-L-R-K-N(638)
Show More Show Less