Learn More
ACROBiosystems Inc ACROBiosystems Mouse OX40 / TNFRSF4 / CD134 Protein, His Tag
Numéro de catalogue. 502106657
Encompass
Recombinant Protein;1MG;Mouse OX40, His Tag (OX0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # P47741-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, Fc Tag (Cat. No. OXL-M526x) at 2 g/mL (100 L/well) can bind Mouse OX40, His Tag (Cat. No. OX0-M5228) with a linear range of 3.9-31.3 ng/mL (QC tested).
Numéro de catalogue. 502106657 Fournisseur ACROBiosystems Inc Code fournisseur OX0M52281MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;1MG;Mouse OX40, His Tag (OX0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # P47741-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, Fc Tag (Cat. No. OXL-M526x) at 2 g/mL (100 L/well) can bind Mouse OX40, His Tag (Cat. No. OX0-M5228) with a linear range of 3.9-31.3 ng/mL (QC tested).