Learn More
Biomatik Corporation Mouse Mucin-5 subtype AC, MUC5AC ELISA Kit, 96 tests
Catalog No. 501498596
Encompass
Mouse Mucin-5 Subtype Ac, Muc5Ac Elisa Kit for detecting Mucin-5 Subtype Ac, Muc5Ac in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.688-300ng/ml. Sensitivity: 1.172ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501498596 Supplier Biomatik Corporation Supplier No. EKC37413
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mouse Mucin-5 Subtype Ac, Muc5Ac Elisa Kit for detecting Mucin-5 Subtype Ac, Muc5Ac in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.688-300ng/ml. Sensitivity: 1.172ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.