missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IgG3 Isotype Control, FITC

Catalog No. MG301
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MG301 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MG301 Supplier Invitrogen™ Supplier No. MG301

Mouse Monoclonal Isotype Control

Mouse IgG3 Isotype Control for Flow

Mouse IgG3 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the least abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG3 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG3 is less commonly used in research and diagnostic applications compared to other IgG subtypes, but it is still used in some applications such as ELISA and immunohistochemistry.

Specifications

Antigen Mouse IgG3
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
Formulation PBS with 0.1% sodium azide
Gene Alias IgG3; Ighg3; Immunoglobulin G; Immunoglobulin G3; Immunoglobulin heavy constant gamma 3; ImmunoglobulinG; ImmunoglobulinG3
Host Species Mouse
Purification Method Purified
Quantity 500 μL
Regulatory Status ASR
Whole Molecule Whole Antibody
Target Species Not Applicable
Content And Storage 2-8°C
Product Type Isotype Control
Form Liquid
Isotype IgG3
No. of Reactions 50 Tests
Shipping Condition Wet Ice
Show More Show Less