missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Mouse IgG2b, APC, Clone: MPC-11, Negative Control, EMD Millpore™

Catalog No. MABF1776MI
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
APC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1776MI 100 μg APC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1776MI Supplier MilliporeSigma™ Supplier No. MABF1776

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects IgG2b Clone: MPC-11 in samples, and it is validated for Flow Cytometry

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Specifications

Antigen IgG2b
Applications Flow Cytometry
Classification Monoclonal
Clone MPC-11
Conjugate APC
Formulation Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. This Monoclonal was purified from tissue culture supernatant via affinity chromatography.
Host Species Mouse
Immunogen Proprietary immunogen
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Test Specificity Mouse IgG2b
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Isotype IgG2b
Show More Show Less

For Research Use Only