missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IgG2a Isotype Control, Pacific Orange™

Catalog No. MG2A30
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MG2A30 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MG2A30 Supplier Invitrogen™ Supplier No. MG2A30

Mouse Monoclonal Isotype Control

Mouse IgG2a Isotype Control for Flow

Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.

Specifications

Antigen Mouse IgG2a
Applications Flow Cytometry
Classification Monoclonal
Conjugate Pacific Orange
Formulation PBS with 0.1% sodium azide
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Mouse
Purification Method Purified
Quantity 500 μL
Regulatory Status ASR
Target Species Not Applicable
Content And Storage 2-8°C
Product Type Isotype Control
Form Liquid
Isotype IgG2a
Shipping Condition Wet Ice
Product Line Pacific Orange
Show More Show Less