missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG2a Isotype Control, APC
Mouse Monoclonal Isotype Control
Supplier: Invitrogen™ MG2A05
Description
Mouse IgG2a Isotype Control for Flow
Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Chemical Identifiers
| Mouse IgG2a | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| ASR | |
| PBS with 0.1% sodium azide | |
| Isotype Control (Primary) | |
| Purified |
| 2-8°C | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Not Applicable | |
| IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
| Isotype Control |
Specifications
| Mouse IgG2a | |
| Monoclonal | |
| PBS with 0.1% sodium azide | |
| Mouse | |
| 500 μL | |
| Whole Antibody | |
| Not Applicable | |
| Isotype Control | |
| IgG2a | |
| Wet Ice |
| Flow Cytometry | |
| APC | |
| IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
| Purified | |
| ASR | |
| Isotype Control (Primary) | |
| 2-8°C | |
| Liquid | |
| 50 Tests |