missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG1 Isotype Control, PerCP
Mouse Monoclonal Isotype Control
Supplier: Invitrogen™ MG131
Description
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Chemical Identifiers
| Mouse IgG1 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| ASR | |
| PBS with 0.1% sodium azide | |
| Isotype Control |
| 2-8°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Not Applicable | |
| IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
| Purified |
Specifications
| Mouse IgG1 | |
| Monoclonal | |
| PBS with 0.1% sodium azide | |
| Mouse | |
| 500 μL | |
| Whole Antibody | |
| 2-8°C | |
| Liquid | |
| 488⁄678 | |
| 50 Tests |
| Flow Cytometry | |
| PerCP | |
| IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
| Purified | |
| ASR | |
| Not Applicable | |
| Isotype Control | |
| IgG1 | |
| Flow Cytometer | |
| Wet Ice |