missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IgG1 Isotype Control Cocktail, FITC, PE

Catalog No. PIPA533180
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533180 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533180 Supplier Invitrogen™ Supplier No. PA533180

Isotype Control Cocktail

PA5-33180 is suitable for use as a negative control for the measurement of non-specific binding of mouse monoclonals of isotype IgG1 in human tissues in a dual labelling technique using FITC and R-Phycoerythrin fluorochromes. This antibody contains a monoclonal mixture of FITC-conjugated Mouse IgG1 RPE-conjugated Mouse IgG1. Reconstitute with 0.5 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.

Specifications

Antigen Mouse IgG1
Applications Control, Flow Cytometry
Classification Cocktail
Concentration Conc. Not Determined
Conjugate FITC/PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene Alias IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Host Species Mouse
Immunogen Antigen not available at this time
Purification Method Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Lyophilized
Isotype IgG1
Show More Show Less