missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse GnRH1 Synthetic Peptide

Catalog No. PEP168
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP168 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP168 Supplier Invitrogen™ Supplier No. PEP168

Synthetic Peptide

Gonadotropin Releasing Hormone (GnRH) Synthetic Peptide, PEP-168, from Invitrogen.

The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).

Specifications

Gene ID (Entrez) 14714
Species Mouse
Purification Method purified
Accession Number P13562
Common Name Mouse GnRH1
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Gnrh1
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence pyroE-H-W-S-Y-G-L-R-P-(G-NH2)
Show More Show Less