Learn More
Kent Scientific Mouse Endotracheal Intubation Kit
Catalog No. 13005167
Encompass
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
13005167 Mouse
1 options
Catalog No. 13005167 Supplier Kent Scientific Supplier No. ETIMSE
Includes
Mouse intubation stand with adjustable head positioning, anesthesia mask, Anesthesia mask tubing and connectors, Fiber optic light and USB connection, trans-tracheal illuminator, 5-pack 20G catheters, suture, dental spatula, dental mirror, power supply
Integrated anesthesia mask allows delivery of isoflurane before intubation.
  • Adjustable neck angle allows aligning airway structures for intubation
  • Fiber optic guide light illuminutes airway to enhance visualization of airway
Specifications
Animal Mouse
For Use With (Application) Intubation of Mice
Type Mouse Endotracheal Intubation Kit
Includes Mouse intubation stand with adjustable head positioning, anesthesia mask, Anesthesia mask tubing and connectors, Fiber optic light and USB connection, trans-tracheal illuminator, 5-pack 20G catheters, suture, dental spatula, dental mirror, power supply