Learn More
Invitrogen™ eBioscience™ Mouse Cytokine Positive Control Cells
Catalog No. 501122024
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
501122024 1 mL
1 options
Catalog No. 501122024 Supplier Invitrogen™ Supplier No. 00450051
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recommended for use as positive control cells when establishing an intracellular cytokine staining protocol.
  • Mouse splenocytes were stimulated with ConA (3μg/mL) for two days, followed by recombinant IL-2 (20ng/mL) and IL-4 (20ng/mL) for three days
  • The cells were then restimulated with immobilized anti-mouse CD3 (10μg/mL) and soluble anti-mouse CD28 (2μg/mL) in the presence of Brefeldin A for 5 hours
  • These cells contain subpopulations that express IL-2, IL-3, IL-4, IL-10, GM-CSF, and IFNγ
  • The cells have been fixed with formaldehyde and are provided in a stabilizing buffer so that they may be stored at −80°C for up to six months or at 2 to 8°C for up to one month
Specifications
For Use With (Equipment) Flow Cytometer
Quantity 1 mL
Product Line eBioscience
Product Type Mouse Cytokine Positive Control Cells
For Research Use Only. Not for use in diagnostic procedures.