Learn More
ACROBiosystems Inc ACROBiosystems Mouse CD39 / ENTPD1 Protein, His Tag
Catalog No. 502106415
Encompass
Recombinant Protein;500UG;Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.7 kDa.
Catalog No. 502106415 Supplier ACROBiosystems Inc Supplier No. CD9M52H3500UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;500UG;Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.7 kDa.