Learn More
ACROBiosystems Inc ACROBiosystems Mouse CD36 / SR-B3 Protein, His Tag
Catalog No. 502107109
Encompass
Recombinant Protein;100UG;Mouse CD36, His Tag (CD6-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Lys 439 (Accession # Q08857-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.3 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 502107109 Supplier ACROBiosystems Inc Supplier No. CD6M52H3100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Mouse CD36, His Tag (CD6-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Lys 439 (Accession # Q08857-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.3 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.