missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, TCI America™

Catalog No. p-7132186
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M046525ML 25 mL
M0465500ML 500 mL
2 options

Catalog No. M046525ML

Supplier: TCI America M046525ML

Only null left

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Melting Point -5°C
Color Yellow
pH 11 (25% H2O soln.)
Boiling Point 129°C
Quantity 25 mL
UN Number 2054
Formula Weight 87.12
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Morpholine
EINECSNumber (5)-0859
RTECSNumber QD6475000
TSCA Yes